Product Details
- SNP ID
-
rs116297522
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:86098931 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAGTGCAAATCTTCAGGGGCCGTCC[A/G]GGACTACAGAGCTGTTTCACCCTAC
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ATP6V0D2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74667635] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ATP6V0D2
- Gene Name
- ATPase H+ transporting V0 subunit d2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_152565.1 |
22 |
UTR 5 |
|
|
NP_689778.1 |
View Full Product Details