Product Details

SNP ID
hCV25961097
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:30677405 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATGCAGGAAGTCCTATAATATCT[C/T]CCATATCTAAAGCATGTTACCACCA
Phenotype
MIM: 603405 MIM: 610990
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DHX16 PubMed Links

Gene Details

Gene
DHX16
Gene Name
DEAH-box helicase 16
There are no transcripts associated with this gene.

Gene
PPP1R18
Gene Name
protein phosphatase 1 regulatory subunit 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134870.1 Intron NP_001128342.1
NM_133471.3 Intron NP_597728.1

View Full Product Details