Product Details

SNP ID
rs142615737
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:79610990 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTAATGCTTGGAGATTCTGACTCT[A/G]CGGGAGTCATGTCAGGGGACCTTGG
Phenotype
MIM: 600621
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
STMN2 PubMed Links

Gene Details

Gene
STMN2
Gene Name
stathmin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199214.1 177 UTR 5 NP_001186143.1
NM_007029.3 177 UTR 5 NP_008960.2
XM_005251142.2 177 Intron XP_005251199.1

View Full Product Details