Product Details

SNP ID
rs116257381
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:98624852 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTTGTGGGTCTAGCAAGATGTGG[C/T]TCTGGGCCGTGCTGGTGGGTGGTGG
Phenotype
MIM: 600750
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NPTX2 PubMed Links

Gene Details

Gene
NPTX2
Gene Name
neuronal pentraxin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002523.2 Intron NP_002514.1

View Full Product Details