Product Details

SNP ID
rs34570972
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:31562121 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACAAAGAAGAACACAAATCATGTC[C/T]CGAGGACTTCCAAAGCAGAAACCGA
Phenotype
MIM: 613621
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NUBPL PubMed Links

Gene Details

Gene
NUBPL
Gene Name
nucleotide binding protein like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001201573.1 203 Intron NP_001188502.1
NM_001201574.1 203 Intron NP_001188503.1
NM_025152.2 203 Silent Mutation TCC,TCT S54S NP_079428.2
XM_011537181.2 203 Intron XP_011535483.1
XM_011537182.2 203 UTR 5 XP_011535484.1
XM_011537183.2 203 Silent Mutation TCC,TCT S54S XP_011535485.1
XM_011537184.2 203 Intron XP_011535486.1
XM_017021664.1 203 Silent Mutation TCC,TCT S54S XP_016877153.1
XM_017021665.1 203 Silent Mutation TCC,TCT S54S XP_016877154.1
XM_017021666.1 203 Silent Mutation TCC,TCT S54S XP_016877155.1
XM_017021667.1 203 UTR 5 XP_016877156.1

View Full Product Details