Product Details

SNP ID
rs10991359
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:104694482 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCAATTTACCTGAATCATGTCCTT[C/T]TCTATACTACCCAGCAGGCAGGTGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR13D1 PubMed Links

Gene Details

Gene
OR13D1
Gene Name
olfactory receptor family 13 subfamily D member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004484.1 61 Missense Mutation CTC,TTC L21F NP_001004484.1

View Full Product Details