Product Details

SNP ID
rs16893967
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:82172386 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TACTAAGTATGTTATTTACCTGAAT[C/T]AGAGCCAACGGTTTTTCTCGACTTC
Phenotype
MIM: 606457
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IBTK PubMed Links

Gene Details

Gene
IBTK
Gene Name
inhibitor of Bruton tyrosine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300906.1 5137 Silent Mutation CTA,CTG L1293L NP_001287835.1
NM_015525.3 5137 Silent Mutation CTA,CTG L1308L NP_056340.2
XM_006715453.2 5137 Silent Mutation CTA,CTG L1349L XP_006715516.1
XM_006715454.2 5137 Silent Mutation CTA,CTG L1334L XP_006715517.1

View Full Product Details