Product Details

SNP ID
rs61745777
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:484057 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCCAGAGGTCCAAGGGGTGAAGG[A/G]GTTGGCACTGAAGAAATCCCTGAAA
Phenotype
MIM: 115440
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CSNK2A1 PubMed Links

Gene Details

Gene
CSNK2A1
Gene Name
casein kinase 2 alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001895.3 1356 Silent Mutation NP_001886.1
NM_177559.2 1356 Silent Mutation NP_808227.1
NM_177560.2 1356 Silent Mutation NP_808228.1

View Full Product Details