Product Details

SNP ID
rs8192593
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:53507321 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAAGTATCAGAACCTAGAGCACAA[A/G]TCTCAACCGAAGTAGAGGCAATGGT
Phenotype
MIM: 606371 MIM: 600447 MIM: 604643 MIM: 605053
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ATF7 PubMed Links

Gene Details

Gene
ATF7
Gene Name
activating transcription factor 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130060.1 318 Intron NP_001123532.1
NM_001206682.1 318 Intron NP_001193611.1
NM_001206683.1 318 Intron NP_001193612.1
NM_006856.2 318 Intron NP_006847.1
XM_005268587.3 318 Intron XP_005268644.1
XM_017018722.1 318 Intron XP_016874211.1
Gene
LOC100652999
Gene Name
uncharacterized LOC100652999
There are no transcripts associated with this gene.

Gene
MAP3K12
Gene Name
mitogen-activated protein kinase kinase kinase 12
There are no transcripts associated with this gene.

Gene
NPFF
Gene Name
neuropeptide FF-amide peptide precursor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320296.1 318 UTR 5 NP_001307225.1
NM_003717.3 318 Intron NP_003708.1
Gene
TARBP2
Gene Name
TARBP2, RISC loading complex RNA binding subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004178.4 318 Intron NP_004169.3
NM_134323.1 318 Intron NP_599150.1
NM_134324.2 318 Intron NP_599151.2
XM_005269114.1 318 Intron XP_005269171.1
XM_005269115.2 318 Intron XP_005269172.1
XM_005269117.1 318 Intron XP_005269174.1
XM_005269120.4 318 Intron XP_005269177.1
XM_005269122.3 318 Intron XP_005269179.1
XM_006719581.1 318 Intron XP_006719644.1
XM_011538712.2 318 Intron XP_011537014.1
XM_017019910.1 318 Intron XP_016875399.1
XM_017019911.1 318 Intron XP_016875400.1

View Full Product Details