Product Details

SNP ID
rs4074320
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:50376086 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCACTGTCTTGGCCGTAAAATATAT[C/T]ATTCAGGATGTTAGCGCTGGCATTC
Phenotype
MIM: 300837
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DGKK PubMed Links

Gene Details

Gene
DGKK
Gene Name
diacylglycerol kinase kappa
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013742.3 3524 Missense Mutation AAT,GAT N1118D NP_001013764.1
XM_017029268.1 3524 Missense Mutation AAT,GAT N1118D XP_016884757.1

View Full Product Details