Product Details

SNP ID
hCV25968846
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:27256296 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAACAGAGACAGATCAAAAAAGA[C/G]TGAAACAATTCCAGTAACTCAAGCT
Phenotype
MIM: 602878
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SLC30A3 PubMed Links

Gene Details

Gene
SLC30A3
Gene Name
solute carrier family 30 member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318949.1 Intron NP_001305878.1
NM_001318950.1 Intron NP_001305879.1
NM_001318951.1 Intron NP_001305880.1
NM_003459.4 Intron NP_003450.2

View Full Product Details