Product Details

SNP ID
rs189623978
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:50375937 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGCCATATTTGCCCAGTTTCTCC[A/G]TCCAGCCTCCGGCTCCACTTCTTTC
Phenotype
MIM: 300837
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DGKK PubMed Links

Gene Details

Gene
DGKK
Gene Name
diacylglycerol kinase kappa
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013742.3 Intron NP_001013764.1
XM_017029268.1 Intron XP_016884757.1

View Full Product Details