Product Details

SNP ID
rs74164152
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:133350271 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTCTGGCTTCAAACCCCAAGCTG[A/G]CTTAGGGGAGAGAGGCATGGGTACG
Phenotype
MIM: 609776
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FUOM PubMed Links

Gene Details

Gene
FUOM
Gene Name
fucose mutarotase
There are no transcripts associated with this gene.

Gene
PRAP1
Gene Name
proline rich acidic protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145201.1 Intron NP_001138673.1
NM_145202.4 Intron NP_660203.3

View Full Product Details