Product Details

SNP ID
rs3213986
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:38880700 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGTCTACACAGGAAGCTGGAAAGA[C/T]GACAAGGTATTATTGTTGTTTTTAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DHX57 PubMed Links

Gene Details

Gene
DHX57
Gene Name
DEAH-box helicase 57
There are no transcripts associated with this gene.

Gene
MORN2
Gene Name
MORN repeat containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145450.1 301 UTR 5 NP_001138922.1

View Full Product Details