Product Details
- SNP ID
-
rs61734985
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:49893950 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTGGGCGGCCAGGACAAGATCCGG[C/G]CGCTCTGGCGGCATTACTACACTGG
- Phenotype
-
MIM: 600464
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ARF6
PubMed Links
Gene Details
- Gene
- ARF6
- Gene Name
- ADP ribosylation factor 6
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001663.3 |
835 |
Missense Mutation |
CCG,GCG |
P72A |
NP_001654.1 |
View Full Product Details