Product Details

SNP ID
rs61736780
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:38880609 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCTCCTCTGTTTAGATGGTGACT[A/G]TACAAGAACATCTTCTGGAATCTAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DHX57 PubMed Links

Gene Details

Gene
DHX57
Gene Name
DEAH-box helicase 57
There are no transcripts associated with this gene.

Gene
MORN2
Gene Name
MORN repeat containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145450.1 210 UTR 5 NP_001138922.1

View Full Product Details