Product Details

SNP ID
rs200158058
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:66963416 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCTGGTGGAGGCGGGCAAACAGG[A/C]AGGTTGCAGGAGAATCCTGCTGCTG
Phenotype
MIM: 605279
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CES3 PubMed Links

Gene Details

Gene
CES3
Gene Name
carboxylesterase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001185176.1 Intron NP_001172105.1
NM_001185177.1 Intron NP_001172106.1
NM_024922.5 Intron NP_079198.2
XM_011522975.2 Intron XP_011521277.1
XM_011522976.2 Intron XP_011521278.1

View Full Product Details