Product Details

SNP ID
rs72625170
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:34360060 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATACCAAAATAGGCCTCCTGGAG[C/T]GGGGTGAGCATAGCCACTAAGCACC
Phenotype
MIM: 612039 MIM: 608963
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LPCAT4 PubMed Links

Gene Details

Gene
LPCAT4
Gene Name
lysophosphatidylcholine acyltransferase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153613.2 Intron NP_705841.2
XM_006720454.2 Intron XP_006720517.1
XM_011521422.1 Intron XP_011519724.1
Gene
NUTM1
Gene Name
NUT midline carcinoma family member 1
There are no transcripts associated with this gene.

View Full Product Details