Product Details

SNP ID
rs34120878
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:500405 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTTCAAACACTTAGGAGCCAGTT[A/C]TAACTGGACCACGCTGCCACCACCT
Phenotype
MIM: 616918
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PIGG PubMed Links

Gene Details

Gene
PIGG
Gene Name
phosphatidylinositol glycan anchor biosynthesis class G
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127178.2 534 Missense Mutation TAT,TCT Y55S NP_001120650.1
NM_001289051.1 534 UTR 5 NP_001275980.1
NM_001289052.1 534 Missense Mutation TAT,TCT Y55S NP_001275981.1
NM_001289053.1 534 UTR 5 NP_001275982.1
NM_001289055.1 534 UTR 5 NP_001275984.1
NM_001289057.1 534 UTR 5 NP_001275986.1
NM_017733.4 534 Missense Mutation TAT,TCT Y55S NP_060203.3
XM_005272284.2 534 UTR 5 XP_005272341.1
XM_011513490.2 534 Missense Mutation TAT,TCT Y55S XP_011511792.1
XM_011513491.1 534 UTR 5 XP_011511793.1
XM_011513492.1 534 Intron XP_011511794.1
XM_011513493.1 534 Missense Mutation TAT,TCT Y55S XP_011511795.1
XM_011513494.2 534 Missense Mutation TAT,TCT Y55S XP_011511796.1
XM_017008321.1 534 UTR 5 XP_016863810.1
XM_017008322.1 534 UTR 5 XP_016863811.1
XM_017008323.1 534 UTR 5 XP_016863812.1
XM_017008324.1 534 UTR 5 XP_016863813.1
XM_017008325.1 534 UTR 5 XP_016863814.1
XM_017008326.1 534 Missense Mutation TAT,TCT Y55S XP_016863815.1
Gene
ZNF721
Gene Name
zinc finger protein 721
There are no transcripts associated with this gene.

View Full Product Details