Product Details

SNP ID
rs190928711
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:120605961 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAAAAACAATGTAAACAATATTT[G/T]TTGGCCAGGATTTTTACTGACTTTT
Phenotype
MIM: 300587
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MCTS1 PubMed Links

Gene Details

Gene
MCTS1
Gene Name
MCTS1, re-initiation and release factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001137554.1 Intron NP_001131026.1
NM_014060.2 Intron NP_054779.1

View Full Product Details