Product Details

SNP ID
rs4932171
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:90250870 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGACTTAGAAGCTGGGAGGAGGAA[A/G]AGAAGGCGGAAACGCAGGTAACTAT
Phenotype
MIM: 602293
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CIB1 PubMed Links

Gene Details

Gene
CIB1
Gene Name
calcium and integrin binding 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277764.1 Intron NP_001264693.1
NM_006384.3 Intron NP_006375.2
XM_006720375.1 Intron XP_006720438.1
Gene
GDPGP1
Gene Name
GDP-D-glucose phosphorylase 1
There are no transcripts associated with this gene.

Gene
TTLL13P
Gene Name
tubulin tyrosine ligase like 13, pseudogene
There are no transcripts associated with this gene.

View Full Product Details