Product Details

SNP ID
rs118140194
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:37634944 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTCTAGAATGTACTTCCTATGCT[C/T]AACATAAAATTCGCAAAGGAAGAGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZFP30 PubMed Links

Gene Details

Gene
ZFP30
Gene Name
ZFP30 zinc finger protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320666.1 1919 UTR 3 NP_001307595.1
NM_001320667.1 1919 UTR 3 NP_001307596.1
NM_001320668.1 1919 UTR 3 NP_001307597.1
NM_001320669.1 1919 UTR 3 NP_001307598.1
NM_001320670.1 1919 UTR 3 NP_001307599.1
NM_001320671.1 1919 UTR 3 NP_001307600.1
NM_014898.2 1919 UTR 3 NP_055713.1

View Full Product Details