Product Details

SNP ID
hCV25974401
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:71440038 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACGGCCCGTAGCAGTCCGGTGCTG[C/G]TCTCCGCCCGCGTCCGGCTCGTGGC
Phenotype
MIM: 606667
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LGR5 PubMed Links

Gene Details

Gene
LGR5
Gene Name
leucine rich repeat containing G protein-coupled receptor 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277226.1 269 UTR 5 NP_001264155.1
NM_001277227.1 269 UTR 5 NP_001264156.1
NM_003667.3 269 UTR 5 NP_003658.1

View Full Product Details