Product Details

SNP ID
rs3747178
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:39570851 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCCGGACCCCGGAGCCCCCCATC[C/T]TCCCCGCCAGGCCTGGAGGAGCCTC
Phenotype
MIM: 608230
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CACNA1I PubMed Links

Gene Details

Gene
CACNA1I
Gene Name
calcium voltage-gated channel subunit alpha1 I
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003406.1 99 Silent Mutation TCC,TCT S33S NP_001003406.1
NM_021096.3 99 Silent Mutation TCC,TCT S33S NP_066919.2
XM_017029035.1 99 Intron XP_016884524.1
XM_017029036.1 99 Intron XP_016884525.1
XM_017029037.1 99 Intron XP_016884526.1

View Full Product Details