Product Details

SNP ID
hCV25981812
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.9:92459895 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTATATATATGTTCAAAGATATT[C/T]CATTACTTTATTTAATAACCTACTT
Phenotype
MIM: 608135 MIM: 611505
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ASPN PubMed Links

Gene Details

Gene
ASPN
Gene Name
asporin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193335.1 Intron NP_001180264.1
NM_017680.4 Intron NP_060150.4
Gene
CENPP
Gene Name
centromere protein P
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012267.2 Intron NP_001012267.1
NM_001286969.1 Intron NP_001273898.1
NM_001286971.1 Intron NP_001273900.1

View Full Product Details