Product Details

SNP ID
rs11539080
Assay Type
Functionally tested
NCBI dbSNP Submissions
20
Location
Chr.1:20482821 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACACAACAGACTGCAAGGGGAAG[G/T]TAGAAAAAGCTCAAGCATTTTTTTC
Phenotype
MIM: 614986
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CAMK2N1 PubMed Links

Gene Details

Gene
CAMK2N1
Gene Name
calcium/calmodulin dependent protein kinase II inhibitor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018584.5 1921 UTR 3 NP_061054.2

View Full Product Details