Product Details

SNP ID
rs17261572
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:120626774 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACCAGTTGTATTGGTCTCTATACTT[A/T]TCAAAGGCGTATTTGTAAGCTTTTC
Phenotype
MIM: 300611 MIM: 300587
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
C1GALT1C1 PubMed Links

Gene Details

Gene
C1GALT1C1
Gene Name
C1GALT1 specific chaperone 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001011551.2 632 Missense Mutation GAA,GAT E131D NP_001011551.1
NM_152692.4 632 Missense Mutation GAA,GAT E131D NP_689905.1
Gene
MCTS1
Gene Name
MCTS1, re-initiation and release factor
There are no transcripts associated with this gene.

View Full Product Details