Product Details

SNP ID
rs3738777
Assay Type
Functionally tested
NCBI dbSNP Submissions
49
Location
Chr.1:109294213 on Build GRCh38
Set Membership
JSNP
Context Sequence [VIC/FAM]
TTGCCTCTCTTTACTTACCTTTGTC[A/G]GAGTACCATGCCTTCTTAGGTGTCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MYBPHL PubMed Links

Gene Details

Gene
MYBPHL
Gene Name
myosin binding protein H like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010985.2 3494 UTR 3 NP_001010985.2
NM_001265613.1 3494 UTR 3 NP_001252542.1
XM_017001173.1 3494 UTR 3 XP_016856662.1
XM_017001174.1 3494 UTR 3 XP_016856663.1
XM_017001175.1 3494 UTR 3 XP_016856664.1

View Full Product Details