Product Details

SNP ID
rs73476336
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:83660393 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAGAATACTGAACCAGAGTCAGC[C/T]AAATGTATCACTGACCCTCAATTAT
Phenotype
MIM: 605046
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
UBQLN1 PubMed Links

Gene Details

Gene
UBQLN1
Gene Name
ubiquilin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013438.4 3654 UTR 3 NP_038466.2
NM_053067.2 3654 UTR 3 NP_444295.1
XM_005251948.2 3654 Intron XP_005252005.1

View Full Product Details