Product Details

SNP ID
rs150851637
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:135052620 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATACCGTGGGTCCCGGGTCTCCTCC[C/T]CCAGTCTTCTTTGGTCAACCGCAAG
Phenotype
MIM: 602536
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAP3K19 PubMed Links

Gene Details

Gene
MAP3K19
Gene Name
mitogen-activated protein kinase kinase kinase 19
There are no transcripts associated with this gene.

Gene
RAB3GAP1
Gene Name
RAB3 GTPase activating protein catalytic subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172435.1 Intron NP_001165906.1
NM_012233.2 Intron NP_036365.1
XM_011510823.2 Intron XP_011509125.1
XM_011510825.2 Intron XP_011509127.1

View Full Product Details