Product Details

SNP ID
rs199900757
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:112619916 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCGCCTCTCTCTCTTACCTCCTC[C/G]CCCGACAGGGAACTGGGGCCCGGCC
Phenotype
MIM: 601580
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CAPZA1 PubMed Links

Gene Details

Gene
CAPZA1
Gene Name
capping actin protein of muscle Z-line alpha subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006135.2 923 Intron NP_006126.1
XM_011542225.2 923 Intron XP_011540527.1
XM_017002424.1 923 Intron XP_016857913.1
Gene
ST7L
Gene Name
suppression of tumorigenicity 7 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308264.1 923 Intron NP_001295193.1
NM_017744.4 923 Intron NP_060214.2
NM_138727.3 923 Intron NP_620055.1
NM_138728.2 923 Intron NP_620056.1
NM_138729.3 923 Intron NP_620057.1
XM_005270963.4 923 Intron XP_005271020.1
XM_005270964.3 923 Intron XP_005271021.1
XM_011541627.2 923 Intron XP_011539929.1
XM_011541628.2 923 Intron XP_011539930.1
XM_011541629.2 923 Intron XP_011539931.1
XM_011541630.2 923 UTR 5 XP_011539932.1
XM_011541631.2 923 Intron XP_011539933.1
XM_017001534.1 923 Intron XP_016857023.1
XM_017001535.1 923 Intron XP_016857024.1
XM_017001536.1 923 Intron XP_016857025.1
XM_017001537.1 923 Intron XP_016857026.1
XM_017001538.1 923 Intron XP_016857027.1

View Full Product Details