Product Details

SNP ID
rs34534864
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:126304087 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACCGCCTCCGCGGCAGCATGGCG[A/G]ACGCAGCTCCTCAACTAGGCAAGAG
Phenotype
MIM: 610534 MIM: 606252
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DCPS PubMed Links

Gene Details

Gene
DCPS
Gene Name
decapping enzyme, scavenger
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014026.4 787 Missense Mutation AAC,GAC N3D NP_054745.1
XM_011542778.2 787 Missense Mutation AAC,GAC N3D XP_011541080.1
XM_011542780.2 787 Intron XP_011541082.1
XM_017017612.1 787 UTR 5 XP_016873101.1
Gene
HP11014
Gene Name
uncharacterized HP11014
There are no transcripts associated with this gene.

Gene
TIRAP
Gene Name
TIR domain containing adaptor protein
There are no transcripts associated with this gene.

View Full Product Details