Product Details

SNP ID
rs6659926
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:32681756 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCTTGCCAAGTTTCTGGCACTCT[G/T]GTCTGGTTGGAAGAGTACATCCAAA
Phenotype
MIM: 602923 MIM: 611750
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RBBP4 PubMed Links

Gene Details

Gene
RBBP4
Gene Name
RB binding protein 4, chromatin remodeling factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135255.1 3388 UTR 3 NP_001128727.1
NM_001135256.1 3388 UTR 3 NP_001128728.1
NM_005610.2 3388 UTR 3 NP_005601.1
Gene
SYNC
Gene Name
syncoilin, intermediate filament protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161708.1 3388 UTR 3 NP_001155180.1
NM_030786.2 3388 UTR 3 NP_110413.2

View Full Product Details