Product Details

SNP ID
rs61754894
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:169822261 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCTTCTCCTCCTCCCTTGTCGTT[A/G]TCAGCTTCCTCCACGCCCTGCGCGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRRC34 PubMed Links

Gene Details

Gene
LRRC34
Gene Name
leucine rich repeat containing 34
There are no transcripts associated with this gene.

Gene
LRRIQ4
Gene Name
leucine rich repeats and IQ motif containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080460.1 451 Missense Mutation ATC,GTC I114V NP_001073929.1
XM_005247421.2 451 Missense Mutation ATC,GTC I114V XP_005247478.1
XM_006713613.3 451 Missense Mutation ATC,GTC I114V XP_006713676.1
XM_011512748.1 451 Missense Mutation ATC,GTC I114V XP_011511050.1
XM_011512749.1 451 Missense Mutation ATC,GTC I114V XP_011511051.1

View Full Product Details