Product Details

SNP ID
rs77140077
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:82614187 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTCCACATATCCACTGGCATTAG[G/T]TTCTGAGAGTCTGATGAGGGACATC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LINC00575 PubMed Links

Gene Details

Gene
LINC00575
Gene Name
long intergenic non-protein coding RNA 575
There are no transcripts associated with this gene.

View Full Product Details