Product Details
- SNP ID
-
rs28708714
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:100704146 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTAAAATTCCTTTAATTCCTCTATA[C/G]TTAGTTCCACTTTTCCTTGAAAAAT
- Phenotype
-
MIM: 604679
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MIR7705
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79077472] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MIR7705
- Gene Name
- microRNA 7705
There are no transcripts associated with this gene.
- Gene
- PABPC1
- Gene Name
- poly(A) binding protein cytoplasmic 1
View Full Product Details