Product Details

SNP ID
rs3754822
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:197462590 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGTATCGGGAGTGGAGGATTACAA[G/A]CATTTTGTTCCTTGGTGCAAAAAAT
Phenotype
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
COQ10B PubMed Links

Gene Details

Gene
COQ10B
Gene Name
coenzyme Q10B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320818.1 488 Silent Mutation AAA,AAG K52K NP_001307747.1
NM_001320819.1 488 Silent Mutation AAA,AAG K74K NP_001307748.1
NM_001320820.1 488 Silent Mutation AAA,AAG K59K NP_001307749.1
NM_025147.4 488 Silent Mutation AAA,AAG K102K NP_079423.1

View Full Product Details