Product Details

SNP ID
rs4758168
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5884818 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTGCCATTCTGTGGGCATAACATC[A/G]TACCTCACACATACTGTGAGCACAG
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
OR52E4 PubMed Links

Gene Details

Gene
OR52E4
Gene Name
olfactory receptor family 52 subfamily E member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005165.1 526 Missense Mutation ATA,GTA I176V NP_001005165.1

View Full Product Details