Product Details

SNP ID
rs116471360
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:151706091 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCCAGTCCTTGACAGTGTGGGCC[A/C]AGTCTCCGGCCACTCCCTCCCCACT
Phenotype
MIM: 612678 MIM: 611541
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CELF3 PubMed Links

Gene Details

Gene
CELF3
Gene Name
CUGBP, Elav-like family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172648.3 Intron NP_001166119.1
NM_001172649.3 Intron NP_001166120.1
NM_001291106.1 Intron NP_001278035.1
NM_001291107.1 Intron NP_001278036.1
NM_007185.6 Intron NP_009116.3
XM_006711139.2 Intron XP_006711202.1
XM_006711141.2 Intron XP_006711204.1
XM_011509111.2 Intron XP_011507413.1
XM_011509112.2 Intron XP_011507414.1
XM_011509114.2 Intron XP_011507416.1
XM_017000145.1 Intron XP_016855634.1
XM_017000146.1 Intron XP_016855635.1
Gene
SNX27
Gene Name
sorting nexin family member 27
There are no transcripts associated with this gene.

View Full Product Details