Product Details

SNP ID
hCV25989732
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:3543398 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCATTGATTTCTTTGGGCTAAAG[A/C]CCATAGATGGAGACTGGAATACTTA
Phenotype
MIM: 616787 MIM: 615648
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CLUAP1 PubMed Links

Gene Details

Gene
CLUAP1
Gene Name
clusterin associated protein 1
There are no transcripts associated with this gene.

Gene
LOC101929732
Gene Name
uncharacterized LOC101929732
There are no transcripts associated with this gene.

Gene
NLRC3
Gene Name
NLR family CARD domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178844.3 Intron NP_849172.2

View Full Product Details