Product Details

SNP ID
rs76311816
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:133928649 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAGCTAGGAATGCCTTCCTGAGC[C/G]TGGTTGGTGGGGGCTCTAAGGCCAT
Phenotype
MIM: 601460
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C3orf36 PubMed Links

Gene Details

Gene
C3orf36
Gene Name
chromosome 3 open reading frame 36
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025041.2 1164 Missense Mutation ACG,AGG T52R NP_079317.2
Gene
SLCO2A1
Gene Name
solute carrier organic anion transporter family member 2A1
There are no transcripts associated with this gene.

View Full Product Details