Product Details

SNP ID
rs16854916
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:108908177 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAGGAGATCCTGATCTTTTGCCA[C/T]GCCATTGATAAATTCTTCTAAAGTC
Phenotype
MIM: 605128
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GUCA1C PubMed Links

Gene Details

Gene
GUCA1C
Gene Name
guanylate cyclase activator 1C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005459.3 543 Missense Mutation ATG,GTG M159V NP_005450.3
XM_005247910.3 543 Missense Mutation CAT,CGT H172R XP_005247967.1
XM_011513334.2 543 Missense Mutation ATG,GTG M75V XP_011511636.1

View Full Product Details