Product Details

SNP ID
rs35245221
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:61366177 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGGACCATCATCCTGACAGCTGT[A/G]TACTTTGCCCTCAGCATCTCCCTTC
Phenotype
MIM: 614459
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CYB561A3 PubMed Links

Gene Details

Gene
CYB561A3
Gene Name
cytochrome b561 family member A3
There are no transcripts associated with this gene.

Gene
TMEM138
Gene Name
transmembrane protein 138
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016464.4 460 Silent Mutation GTA,GTG V87V NP_057548.1
XM_006718585.3 460 Silent Mutation GTA,GTG V87V XP_006718648.1
XM_006718586.2 460 Silent Mutation GTA,GTG V87V XP_006718649.1
XM_006718588.3 460 Silent Mutation GTA,GTG V29V XP_006718651.1
XM_011545098.2 460 Silent Mutation GTA,GTG V87V XP_011543400.1
XM_011545099.1 460 Silent Mutation GTA,GTG V87V XP_011543401.1
XM_011545100.1 460 Silent Mutation GTA,GTG V87V XP_011543402.1
XM_017017917.1 460 Silent Mutation GTA,GTG V87V XP_016873406.1

View Full Product Details