Product Details
- SNP ID
-
rs61745589
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:39098287 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCTGTCCAGCCCCTCAGCCTCCGT[A/C]CCAGTATCTAGGCCCTCTGTGCTTG
- Phenotype
-
MIM: 606867
MIM: 611430
MIM: 612167
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
GORASP1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1109643] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GORASP1
- Gene Name
- golgi reassembly stacking protein 1
- Gene
- TTC21A
- Gene Name
- tetratricopeptide repeat domain 21A
There are no transcripts associated with this gene.
- Gene
- WDR48
- Gene Name
- WD repeat domain 48
There are no transcripts associated with this gene.
View Full Product Details