Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001195010.1 | 1529 | Intron | NP_001181939.1 | ||
NM_021180.3 | 1529 | Intron | NP_067003.2 | ||
NM_198173.2 | 1529 | Intron | NP_937816.1 | ||
NM_198174.2 | 1529 | Intron | NP_937817.3 | ||
XM_011541869.1 | 1529 | Intron | XP_011540171.1 | ||
XM_011541870.2 | 1529 | Intron | XP_011540172.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001199012.1 | 1529 | UTR 3 | NP_001185941.1 | ||
NM_001199013.1 | 1529 | UTR 3 | NP_001185942.1 | ||
NM_001199014.1 | 1529 | UTR 3 | NP_001185943.1 | ||
NM_178122.4 | 1529 | UTR 3 | NP_835223.1 | ||
XM_006711025.2 | 1529 | UTR 3 | XP_006711088.1 | ||
XM_011542403.2 | 1529 | UTR 3 | XP_011540705.1 | ||
XM_011542404.1 | 1529 | UTR 3 | XP_011540706.1 | ||
XM_017002768.1 | 1529 | UTR 3 | XP_016858257.1 | ||
XM_017002769.1 | 1529 | UTR 3 | XP_016858258.1 | ||
XM_017002770.1 | 1529 | UTR 3 | XP_016858259.1 |