Product Details

SNP ID
rs12119046
Assay Type
Functionally tested
NCBI dbSNP Submissions
48
Location
Chr.1:68477982 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCCAGGATTTTCAATCTGTAGTGA[G/T]CAAAATGATATAACTCTGTTAATTC
Phenotype
MIM: 612002
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DEPDC1 PubMed Links

Gene Details

Gene
DEPDC1
Gene Name
DEP domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114120.2 Intron NP_001107592.1
NM_017779.5 Intron NP_060249.2

View Full Product Details