Product Details

SNP ID
rs3093588
Assay Type
Functionally tested
NCBI dbSNP Submissions
34
Location
Chr.1:25343165 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGCATAGATTTAAAATTCTAAATA[C/T]TATCAGATTAACACACTGGACATGA
Phenotype
MIM: 605348
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RSRP1 PubMed Links

Gene Details

Gene
RSRP1
Gene Name
arginine and serine rich protein 1
There are no transcripts associated with this gene.

Gene
TMEM50A
Gene Name
transmembrane protein 50A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014313.3 Intron NP_055128.1
XM_005245817.1 Intron XP_005245874.1
XM_011541159.1 Intron XP_011539461.1

View Full Product Details