Product Details

SNP ID
rs61744665
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:1434527 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTTTATGAGGGACGCCTTCCAGC[A/G]CAGCCACCTGTCCAGAGATGCGGCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC154 PubMed Links

Gene Details

Gene
CCDC154
Gene Name
coiled-coil domain containing 154
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143980.1 2052 Missense Mutation CGC,TGC R629C NP_001137452.1
Gene
LOC105371045
Gene Name
uncharacterized LOC105371045
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011522780.2 2052 Intron XP_011521082.1

View Full Product Details