Product Details

SNP ID
rs61745561
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:3531979 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCGTATAGCTGAGGCCGAACACTG[C/T]GACATAGGTGTCTGGAGACAGAGAA
Phenotype
MIM: 604123
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RNASEH1 PubMed Links

Gene Details

Gene
RNASEH1
Gene Name
ribonuclease H1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286834.1 Intron NP_001273763.1
NM_001286837.1 Intron NP_001273766.1
NM_002936.4 Intron NP_002927.2

View Full Product Details