Product Details

SNP ID
rs150758797
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:9934663 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGAAAATTGTTAGTTGTGGCTCT[A/G]AAATCACCTGGCAAATTTTTAAGCC
Phenotype
MIM: 610458 MIM: 608700
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LZIC PubMed Links

Gene Details

Gene
LZIC
Gene Name
leucine zipper and CTNNBIP1 domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316973.1 Intron NP_001303902.1
NM_001316974.1 Intron NP_001303903.1
NM_001316975.1 Intron NP_001303904.1
NM_001316976.1 Intron NP_001303905.1
NM_032368.4 Intron NP_115744.2
XM_005263506.2 Intron XP_005263563.1
XM_005263509.2 Intron XP_005263566.1
XM_017002546.1 Intron XP_016858035.1
XM_017002547.1 Intron XP_016858036.1
Gene
NMNAT1
Gene Name
nicotinamide nucleotide adenylyltransferase 1
There are no transcripts associated with this gene.

View Full Product Details